Order Entry
Export
Orders LinkContactUsLinkComponent
5-Nitroisoquinoline

5-Nitroisoquinoline

New Product
Supplier:   Apollo Scientific
5-Nitroisoquinoline
Supplier:  Apollo Scientific
New Product
Certificates
5-Nitroisoquinoline

2 Options Available Below

Product Details & Documents
  • CAS Number:607-32-9
  • Formula:C₉H₆N₂O₂
  • MDL Number:MFCD00006905
  • Molecular Weight:174.16 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 290,00 €
    1 290,00 €
    1 * 2,5 kg
       1 290,00 €

    Specifications:
    New Product
    New Product