2-(Ethylthio)nicotinic acid
Jump to:
About this item
- CAS Number:27868-76-4
- Formula:C₈H₉NO₂S
- MDL Number:MFCD00051680
- Molecular Weight:183.23 g/mol
- Storage Temperature:Ambient
2 Options Available Below
- CAS Number:27868-76-4
- Formula:C₈H₉NO₂S
- MDL Number:MFCD00051680
- Molecular Weight:183.23 g/mol
- Storage Temperature:Ambient



