Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Hydroxycinnamic acid 98%

Supplier:   Apollo Scientific
4-Hydroxycinnamic acid 98%
Supplier:  Apollo Scientific
Certificates
4-Hydroxycinnamic acid 98%

5 Options Available Below

Product Details & Documents
Synonyms:  p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid
  • CAS Number:7400-08-0
  • Formula:C₉H₈O₃
  • MDL Number:MFCD00004399
  • Molecular Weight:164.16 g/mol
  • Boiling Pt:346 °C
  • Melting Pt:212…214 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 23,20 €
    23,20 €
    1 * 25 g
       23,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 55,00 €
    55,00 €
    1 * 100 g
       55,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 76,70 €
    76,70 €
    1 * 500 g
       76,70 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 375,00 €
    375,00 €
    1 * 2,5 kg
       375,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 1 490,00 €
    1 490,00 €
    1 * 10 kg
       1 490,00 €

    Specifications:
    New Product
    New Product