Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dichloropyridazine
3,6-Dichloropyridazine 96%
3,6-Dichloropyridazine 96%
:
Apollo Scientific
3,6-Dichloropyridazine 96%
:
Apollo Scientific
:
141-30-0
:
:
C₄H₂Cl₂N₂
:
MFCD00006466
:
148.98 g/mol
:
66…70 °C
:
Ambient
:
205-478-6
:
2811
:
6.1,III
:
GHS06