Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4,6-di-tert-butylphenol
2-Bromo-4,6-di-tert-butylphenol 97%
2-Bromo-4,6-di-tert-butylphenol 97%
Irritant
2-Bromo-4,6-di-tert-butylphenol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Bromo-4,6-di-tert-butylphenol 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
20834-61-1
:
Formula:
C₁₄H₂₁BrO
MDL Number:
MFCD00051597
:
285.22 g/mol
Melting Pt:
56…57 °C
:
GHS07
Storage Temperature:
Ambient