
Ethyl-4-amino-3,5-dichlorobenzoate
Catalog # APOSOR21247-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:234.793 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:74878-31-2
Formula:C₉H₉Cl₂NO₂
MDL Number:MFCD00016636