
2,3-Dimethyl-7-nitroindole 95+%
Catalog # APOSOR2112-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:164…165 °C
:190.2 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:41018-86-4
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD00022710