Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Nitrophenyl)imidazo[1,2-a]pyridine
2-(4-Nitrophenyl)imidazo[1,2-a]pyridine
2-(4-Nitrophenyl)imidazo[1,2-a]pyridine
2-(4-Nitrophenyl)imidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
2-(4-Nitrophenyl)imidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
3323-26-0
:
Formula:
C₁₃H₉N₃O₂
MDL Number:
MFCD00276341
:
239.24 g/mol
Storage Temperature:
Ambient