
3-Imidazo[2,1-b]thiazol-6-ylphenylamine
Catalog # APOSOR2104-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.28 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:861206-26-0
Formula:C₁₁H₉N₃S
MDL Number:MFCD03617826