Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dibromo-4-picoline
2,5-Dibromo-4-picoline
2,5-Dibromo-4-picoline
Corrosive
Irritant
2,5-Dibromo-4-picoline
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
2,5-Dibromo-4-picoline
Supplier:
Apollo Scientific
About this item
CAS Number:
3430-26-0
:
Formula:
C₆H₅Br₂N
MDL Number:
MFCD00234955
Melting Pt:
36…42 °C
:
250.92 g/mol
:
GHS05
:
GHS07
Storage Temperature:
Ambient