Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 3,5-dichlorobenzoate
Ethyl-3,5-dichlorobenzoate 97%
Ethyl-3,5-dichlorobenzoate 97%
:
Apollo Scientific
Ethyl-3,5-dichlorobenzoate 97%
:
Apollo Scientific
:
91085-56-2
:
:
C₉H₈Cl₂O₂
:
MFCD00221467
:
219.07 g/mol
:
35…36 °C
:
GHS07
:
Ambient