(S,S)-(+)-2,5-Hexanediol
Catalog # APOSOR20022-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:118.18 g/mol
Boiling Pt:212…215 °C
Melting Pt:50…53 °C
Flash Pt:102 °C
:GHS07
Storage Temperature:Ambient
CAS Number:34338-96-0
Formula:C₆H₁₄O₂
MDL Number:MFCD00082583