
4,5-Diamino-6-hydroxypyrimidine
Catalog # APOSOR200071-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:126.1200 g/mol
Melting Pt:241 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1672-50-0
Formula:C₄H₆N₄O
MDL Number:MFCD00006114