0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,1'-Biphenyl-2,2'-diamine
1,1'-Biphenyl-2,2'-diamine
1,1'-Biphenyl-2,2'-diamine
1,1'-Biphenyl-2,2'-diamine
Supplier:
Apollo Scientific
Certificates
Expand image
1,1'-Biphenyl-2,2'-diamine
Supplier:
Apollo Scientific
About this item
CAS Number:
1454-80-4
:
Formula:
C₁₂H₁₂N₂
MDL Number:
MFCD00034060
:
184.24 g/mol
Melting Pt:
81 °C
Storage Temperature:
Ambient