4-Nitro-1,3-benzoxazol-2(3H)-one
Catalog # APOSOR200021-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.12 g/mol
Melting Pt:227…228 °C
Storage Temperature:Ambient
CAS Number:28955-71-7
Formula:C₇H₄N₂O₄
MDL Number:MFCD00460557