Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4,6-dichloropyridazine-3-carboxylate
Methyl-4,6-dichloropyridazine-3-carboxylate 97%
Methyl-4,6-dichloropyridazine-3-carboxylate 97%
Irritant
Methyl-4,6-dichloropyridazine-3-carboxylate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-4,6-dichloropyridazine-3-carboxylate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
372118-01-9
:
Formula:
C₆H₄Cl₂N₂O₂
MDL Number:
MFCD09953612
:
207.0142 g/mol
:
GHS07
Storage Temperature:
Ambient