Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Imidazo[1,2-a]pyridine-7-carbaldehyde
Imidazo[1,2-a]pyridine-7-carbaldehyde
Imidazo[1,2-a]pyridine-7-carbaldehyde
:
Apollo Scientific
Imidazo[1,2-a]pyridine-7-carbaldehyde
:
Apollo Scientific
:
136117-73-2
:
:
C₈H₆N₂O
:
MFCD09910326
:
146.15 g/mol
:
143…144 °C
:
GHS07
:
Ambient