Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-7-methylimidazo[1,2-a]pyridine
3-Bromo-7-methylimidazo[1,2-a]pyridine
3-Bromo-7-methylimidazo[1,2-a]pyridine
Irritant
3-Bromo-7-methylimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromo-7-methylimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
56051-32-2
:
Formula:
C₈H₇BrN₂
MDL Number:
MFCD08056208
:
211.0600 g/mol
:
GHS07
Storage Temperature:
Ambient