
2,4-Dimethylbenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:611-01-8
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00002480
:150.18 g/mol
Boiling Pt:267 °C
Melting Pt:124…126 °C
Flash Pt:154 °C
Storage Temperature:Refrigerator
EINECS:210-246-2
:GHS07