0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
5,6-Dichloronicotinic acid
5,6-Dichloronicotinic acid 97%
5,6-Dichloronicotinic acid 97%
APOSOR1909-1G
:
Apollo Scientific
:
5,6-Dichloronicotinic acid 97%
APOSOR1909-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
163…168 °C (dec.)
:
192 g/mol
:
GHS07
:
Ambient
:
41667-95-2
:
C₆H₃Cl₂NO₂
:
MFCD00075181