Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2'-Bipyridine-5-carboxylic acid
2,2'-Bipyridine-5-carboxylic acid 95%
2,2'-Bipyridine-5-carboxylic acid 95%
:
Apollo Scientific
2,2'-Bipyridine-5-carboxylic acid 95%
:
Apollo Scientific
:
1970-80-5
:
:
C₁₁H₈N₂O₂
:
MFCD08234850
:
200.2 g/mol
:
196…198 °C
:
Ambient
:
2811
:
6.1,III
:
GHS06