Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Diamino-2-propanol
1,3-Diamino-2-propanol, Technical Grade
1,3-Diamino-2-propanol, Technical Grade
:
Apollo Scientific
1,3-Diamino-2-propanol, Technical Grade
:
Apollo Scientific
:
616-29-5
:
:
C₃H₁₀N₂O
:
MFCD00008142
:
90.13 g/mol
:
235 °C
:
40…45 °C
:
>110 °C
:
Ambient
:
210-474-2
:
GHS07