Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Diacryloylpiperazine
1,4-Diacryloylpiperazine 95%
1,4-Diacryloylpiperazine 95%
Irritant
1,4-Diacryloylpiperazine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,4-Diacryloylpiperazine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
6342-17-2
:
Formula:
C₁₀H₁₄N₂O₂
MDL Number:
MFCD00077661
:
194.23 g/mol
Melting Pt:
91.5…93.5 °C
:
GHS07
Storage Temperature:
Ambient