Order Entry
Export
Orders LinkContactUsLinkComponent
Uridine 99%

Uridine 99%

Irritant
Supplier:   Apollo Scientific
Uridine 99%
Supplier:  Apollo Scientific
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Melting Pt:163…169 °C
  • Storage Temperature:Refrigerator

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 65,20 €
    65,20 €
    1 * 100 g
       65,20 €

  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 152,00 €
    152,00 €
    1 * 250 g
       152,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 290,00 €
    290,00 €
    1 * 500 g
       290,00 €

    Specifications:
    New Product
    New Product