Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethyl-6-hydroxypyrimidine
2,4-Dimethyl-6-hydroxypyrimidine
2,4-Dimethyl-6-hydroxypyrimidine
Irritant
2,4-Dimethyl-6-hydroxypyrimidine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Dimethyl-6-hydroxypyrimidine
Supplier:
Apollo Scientific
About this item
CAS Number:
6622-92-0
:
Formula:
C₆H₈N₂O
MDL Number:
MFCD00006105
:
124.14 g/mol
Melting Pt:
198…200 °C
EINECS:
229-577-9
:
GHS07
Storage Temperature:
Ambient