2,6-Dibromophenol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:608-33-3
:
Formula:C₆H₄Br₂O
MDL Number:MFCD00002152
:251.91 g/mol
Boiling Pt:255…256 °C
Melting Pt:53…57 °C
Storage Temperature:Ambient
EINECS:210-161-0
UN:2811
ADR:6.1,III
:GHS07