ω,2,4-Trichloroacetophenone
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone
- CAS Number:4252-78-2
- Formula:C₈H₅Cl₃O
- MDL Number:MFCD00000934
- Molecular Weight:223.49 g/mol
- Boiling Pt:130…135 °C/4mmHg
- Melting Pt:47…56 °C
- Density:1.312 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:224-218-2
- UN:2923
- ADR:8+6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: 2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone
- CAS Number:4252-78-2
- Formula:C₈H₅Cl₃O
- MDL Number:MFCD00000934
- Molecular Weight:223.49 g/mol
- Boiling Pt:130…135 °C/4mmHg
- Melting Pt:47…56 °C
- Density:1.312 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:224-218-2
- UN:2923
- ADR:8+6.1,III



