Order Entry
Export
ContactUsLinkComponent
 
3-(1,3-Benzodioxol-5-yl)-1H-pyrazole
Catalog # APOSOR183457-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(1,3-Benzodioxol-5-yl)-1H-pyrazole
Catalog # APOSOR183457-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.19 g/mol
Storage Temperature:Ambient
CAS Number:141791-06-2
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD03618498