

1,8-Diaminonaphthalene 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:479-27-6
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004033
:158.2 g/mol
Boiling Pt:205 °C/12mmHg
Melting Pt:62…64 °C
Flash Pt:195 °C
:GHS07
:GHS08
Storage Temperature:Ambient