

α-Chloro-p-xylene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:104-82-5
:
Formula:C₈H₉Cl
MDL Number:MFCD00000919
:140.61 g/mol
Boiling Pt:199…201 °C
Melting Pt:44351 °C
Density:1.06 g/cm³ (25 °C)
Flash Pt:75 °C
Storage Temperature:Refrigerator
EINECS:203-241-1
UN:3265
ADR:8,III
:GHS05
:GHS07