Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Diacetoxybenzene
1,4-Diacetoxybenzene 98%
1,4-Diacetoxybenzene 98%
Irritant
1,4-Diacetoxybenzene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,4-Diacetoxybenzene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1205-91-0
:
Formula:
C₁₀H₁₀O₄
MDL Number:
MFCD00011643
:
194.19 g/mol
Melting Pt:
121…123 °C
EINECS:
214-887-9
:
GHS07
Storage Temperature:
Ambient