Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Boc-L-serine
N-Boc-L-serine
N-Boc-L-serine
N-Boc-L-serine
Supplier:
Apollo Scientific
Certificates
Expand image
N-Boc-L-serine
Supplier:
Apollo Scientific
About this item
CAS Number:
3262-72-4
:
Formula:
C₈H₁₅NO₅
MDL Number:
MFCD00037243
:
205.21 g/mol
Melting Pt:
91 °C
Flash Pt:
186 °C
Storage Temperature:
Refrigerator