Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furo[3,2-b]pyridine
Furo[3,2-b]pyridine
Furo[3,2-b]pyridine
Irritant
Furo[3,2-b]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Furo[3,2-b]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
272-62-8
:
Formula:
C₇H₅NO
MDL Number:
MFCD11870744
:
119.12 g/mol
Boiling Pt:
79…80 °C/12mmHg
:
GHS07
Storage Temperature:
Ambient