2-(4-Bromobenzyl)-1,3-thiazole-4-carboxylic acid
Catalog # APOSOR17895-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:298.16 g/mol
Storage Temperature:Ambient
CAS Number:1086380-12-2
Formula:C₁₁H₈BrNO₂S
MDL Number:MFCD09258846