
4-Bromo-2,6-dimethylpyridine
Catalog # APOSOR17852-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.0491 g/mol
Boiling Pt:81 °C/14mmHg
Melting Pt:32…33 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5093-70-9
Formula:C₇H₈BrN
MDL Number:MFCD00671475