Order Entry
Export
ContactUsLinkComponent
 
3,4-Dimethoxythiophene
Catalog # APOSOR17831-5G
Supplier:  Apollo Scientific
CAS Number:  
3,4-Dimethoxythiophene
Catalog # APOSOR17831-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :144.19 g/mol
Boiling Pt:100…102 °C/10mmHg
Density:1.209 g/cm³ (25 °C)
Flash Pt:106.7 °C
Storage Temperature:Freezer
CAS Number:51792-34-8
Formula:C6H8O2S
MDL Number:MFCD01096546