
Methyl-5-methyl-2-nitrobenzoate
Catalog # APOSOR17822-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:195.1721 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:20587-30-8
Formula:C₉H₉NO₄
MDL Number:MFCD00157275