
N-(4-Aminophenyl)-N-methyl-4,6-dimethyl-2-pyrimidinamine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220844-79-1
:
Formula:C₁₃H₁₆N₄
MDL Number:MFCD00202978
:228.3 g/mol
Melting Pt:134…136 °C
:GHS07
Storage Temperature:Ambient