1,3-Cyclohexanediol 97%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Cyclohexane-1,3-diol
- CAS Number:504-01-8
- Formula:C₆H₁₂O₂
- MDL Number:MFCD00039458
- Molecular Weight:116.16 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: Cyclohexane-1,3-diol
- CAS Number:504-01-8
- Formula:C₆H₁₂O₂
- MDL Number:MFCD00039458
- Molecular Weight:116.16 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



