


2,5-Dichloropyrimidine 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22536-67-0
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00673131
:148.97 g/mol
Melting Pt:48…55 °C
Flash Pt:107 °C
UN:3077
ADR:9+EHS,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient