Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
Irritant
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
Supplier:
Apollo Scientific
About this item
CAS Number:
7355-55-7
:
MFCD09033259
Formula:
C₆H₆N₄O
:
150.14 g/mol
Melting Pt:
323…324 °C
:
GHS07
Storage Temperature:
Ambient