Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichlorophenol

2,4-Dichlorophenol

Bottle, 10 g
Catalog # APOSOR17023-10G
Corrosive
Toxic
Environment
Supplier:   Apollo Scientific
2,4-Dichlorophenol
Catalog # APOSOR17023-10G
Supplier:  Apollo Scientific
CAS Number:  120-83-2
2,4-Dichlorophenol

Bottle, 10 g

CAS Number:  120-83-2
Price
21,80 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene
  • CAS Number:120-83-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002169
  • Molecular Weight:163 g/mol
  • Boiling Pt:209…210 °C
  • Melting Pt:42…43 °C
  • Density:1.38 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:2928
  • ADR:6.1+8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications