1,9-Nonanediol 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1,9-Dihydroxy nonane, Nonane-1,9-diol
- CAS Number:3937-56-2
- Formula:C₉H₂₀O₂
- MDL Number:MFCD00002991
- Molecular Weight:160.26 g/mol
- Boiling Pt:177 °C/15mmHg
- Melting Pt:45…47 °C
- Flash Pt:113 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: 1,9-Dihydroxy nonane, Nonane-1,9-diol
- CAS Number:3937-56-2
- Formula:C₉H₂₀O₂
- MDL Number:MFCD00002991
- Molecular Weight:160.26 g/mol
- Boiling Pt:177 °C/15mmHg
- Melting Pt:45…47 °C
- Flash Pt:113 °C
- Storage Temperature:Ambient



