
3-Chloroimidazo[1,2-a]pyridine-6-carbohydrazide
Catalog # APOSOR16910-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.62 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1211364-66-7
Formula:C₈H₇ClN₄O
MDL Number:MFCD11100124