Order Entry
Export
ContactUsLinkComponent
 
4,6-Dioxo-6-phenylhexanoic acid
Catalog # APOSOR16866-500MG
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dioxo-6-phenylhexanoic acid
Catalog # APOSOR16866-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :220.23 g/mol
Storage Temperature:Ambient
CAS Number:114150-57-1
Formula:C₁₂H₁₂O₄
MDL Number:MFCD12025948