Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-[1,1'-Biphenyl]-4-yl-1-ethanol
2-[1,1'-Biphenyl]-4-yl-1-ethanol
2-[1,1'-Biphenyl]-4-yl-1-ethanol
2-[1,1'-Biphenyl]-4-yl-1-ethanol
APOSOR16854-1G
:
Apollo Scientific
:
2-[1,1'-Biphenyl]-4-yl-1-ethanol
APOSOR16854-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
198.27 g/mol
:
GHS07
:
Ambient
:
37729-18-3
:
C₁₄H₁₄O
:
MFCD09926182