Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methyl-2-phenylacetamide
N-Methyl-2-phenylacetamide
N-Methyl-2-phenylacetamide
Irritant
N-Methyl-2-phenylacetamide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
N-Methyl-2-phenylacetamide
Supplier:
Apollo Scientific
About this item
CAS Number:
6830-82-6
:
Formula:
C₉H₁₁NO
MDL Number:
MFCD00051354
:
149.19 g/mol
Boiling Pt:
330.3 °C
Melting Pt:
53…58 °C
:
GHS07
Storage Temperature:
Ambient