Order Entry
Export
ContactUsLinkComponent
2,6-Dimethylnicotinic acid 98%
Irritant
2,6-Dimethylnicotinic acid 98%
Catalog # APOSOR16447-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dimethylnicotinic acid 98%
Catalog # APOSOR16447-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:168…169 °C
 :151.17 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:5860-71-9
Formula:C₈H₉NO₂
MDL Number:MFCD06738677