Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dichloro-2-methylpropane
1,2-Dichloro-2-methylpropane 97%
1,2-Dichloro-2-methylpropane 97%
:
Apollo Scientific
1,2-Dichloro-2-methylpropane 97%
:
Apollo Scientific
:
594-37-6
:
:
C₄H₈Cl₂
:
MFCD00000817
:
127.01 g/mol
:
105…106 °C
:
1.095 g/cm³ (25 °C)
:
16 °C
:
Freezer
:
209-837-8
:
1992
:
3+6.1,II
:
GHS02
:
GHS07