
4-Chloro-1,3-dioxolan-2-one, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3967-54-2
:
Formula:C₃H₃ClO₃
MDL Number:MFCD00005383
:122.50 g/mol
Boiling Pt:121…123 °C/18mmHg
Density:1.504 g/cm³ (25 °C)
Flash Pt:>113 °C
EINECS:223-583-5
:GHS07
Storage Temperature:Refrigerator